Product Details

SNP ID
rs144008264
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1772513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCGCGCCTCGCTCTCAGTCTTC[C/T]CTGATGAAGAAAAAGAGGCGTCTGC
Phenotype
MIM: 610886 MIM: 605431 MIM: 611994 MIM: 601817 MIM: 611659
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EME2 PubMed Links

Gene Details

Gene
EME2
Gene Name
essential meiotic structure-specific endonuclease subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257370.1 406 Intron NP_001244299.1
XM_017023025.1 406 Intron XP_016878514.1
XM_017023026.1 406 Intron XP_016878515.1
Gene
MAPK8IP3
Gene Name
mitogen-activated protein kinase 8 interacting protein 3
There are no transcripts associated with this gene.

Gene
MRPS34
Gene Name
mitochondrial ribosomal protein S34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300900.1 406 Missense Mutation GAG,GGG E129G NP_001287829.1
NM_023936.1 406 Missense Mutation GAG,GGG E122G NP_076425.1
XM_017023595.1 406 Missense Mutation GAG,GGG E152G XP_016879084.1
Gene
NME3
Gene Name
NME/NM23 nucleoside diphosphate kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002513.2 406 Intron NP_002504.2
XM_005255332.4 406 Intron XP_005255389.1
XM_011522503.2 406 Intron XP_011520805.1
XM_011522504.2 406 Intron XP_011520806.1
Gene
SPSB3
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
There are no transcripts associated with this gene.

View Full Product Details