Product Details

SNP ID
rs144665805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30086838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCCCGCAGTTTCCTCTTCACAC[A/G]GGCGTCTCGCTCCCTGGGGAACAGT
Phenotype
MIM: 602035 MIM: 602427 MIM: 609724
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928595 PubMed Links

Gene Details

Gene
LOC101928595
Gene Name
uncharacterized LOC101928595
There are no transcripts associated with this gene.

Gene
PPP4C
Gene Name
protein phosphatase 4 catalytic subunit
There are no transcripts associated with this gene.

Gene
TBX6
Gene Name
T-box 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004608.3 938 Missense Mutation CGT,TGT R285C NP_004599.2
XM_005255523.2 938 Missense Mutation CGT,TGT R285C XP_005255580.1
XM_011545926.2 938 Missense Mutation CGT,TGT R285C XP_011544228.1
XM_017023614.1 938 Missense Mutation CGT,TGT R285C XP_016879103.1
Gene
YPEL3
Gene Name
yippee like 3
There are no transcripts associated with this gene.

View Full Product Details