Product Details

SNP ID
rs144829486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30700054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGAGATAAAGCAATTATAGGTC[C/T]GTATTGGGGATTTTCACTCTAGAGA
Phenotype
MIM: 611421
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC730183 PubMed Links

Gene Details

Gene
LOC730183
Gene Name
uncharacterized LOC730183
There are no transcripts associated with this gene.

Gene
SRCAP
Gene Name
Snf2-related CREBBP activator protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006662.2 Intron NP_006653.2

View Full Product Details