Product Details

SNP ID
rs145121462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57977984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGCGCCACATGGACAGGGACAC[C/T]GTCAAGGCCTGCCTTCCTGATGAGC
Phenotype
MIM: 600724 MIM: 610264
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNGB1 PubMed Links

Gene Details

Gene
CNGB1
Gene Name
cyclic nucleotide gated channel beta 1
There are no transcripts associated with this gene.

Gene
TEPP
Gene Name
testis, prostate and placenta expressed
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199046.2 370 Silent Mutation ACC,ACT T111T NP_950247.2
NM_199456.2 370 Silent Mutation ACC,ACT T111T NP_955535.2
XM_017023218.1 370 Silent Mutation ACC,ACT T56T XP_016878707.1

View Full Product Details