Product Details
- SNP ID
-
rs145327184
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:23755877 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGATCTCCAGCAGATAGGGGCGCT[C/T]GCCGTGAACCCCCTGGGAGACCGAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CHP2
PubMed Links
Gene Details
- Gene
- CHP2
- Gene Name
- calcineurin like EF-hand protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022097.3 |
594 |
Silent Mutation |
CTC,CTT |
L57L |
NP_071380.1 |
View Full Product Details