Product Details
- SNP ID
-
rs145408554
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:66378259 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGAAGGCTTTAAATCTTACCCCAT[A/G]GCAGCTGCTGGGACAAGGGAAGTCA
- Phenotype
-
MIM: 601120
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDH5
PubMed Links
Gene Details
- Gene
- CDH5
- Gene Name
- cadherin 5
View Full Product Details