Product Details
- SNP ID
-
rs145842898
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:56565116 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCGTGACAGCACTGGAGCCTTTC[A/G]GACACCTGGACCATGGACCCCAGGG
- Phenotype
-
MIM: 606206
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MT4
PubMed Links
Gene Details
- Gene
- MT4
- Gene Name
- metallothionein 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032935.2 |
68 |
UTR 5 |
|
|
NP_116324.1 |
View Full Product Details