Product Details

SNP ID
rs146060040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30699482 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAAGGCGCAGGTATGGGATATA[C/T]AGCGCCCTGCCCTGAATCCTGGGCG
Phenotype
MIM: 611421
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC730183 PubMed Links

Gene Details

Gene
LOC730183
Gene Name
uncharacterized LOC730183
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256932.1 Intron NP_001243861.1
Gene
SRCAP
Gene Name
Snf2-related CREBBP activator protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006662.2 Intron NP_006653.2

View Full Product Details