Product Details

SNP ID
rs146846062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:55326542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCGCTGCAGCTGCCCAGAGCTA[C/G]CCTGGCTACCTGCCCTATAGCCCAG
Phenotype
MIM: 606196
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IRX6 PubMed Links

Gene Details

Gene
IRX6
Gene Name
iroquois homeobox 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024335.2 785 Nonsense Mutation TAC,TAG Y84* NP_077311.2
XM_005256137.2 785 Nonsense Mutation TAC,TAG Y84* XP_005256194.1

View Full Product Details