Product Details

SNP ID
rs146909074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:798717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGTGAGAGGGGCCGGGTGCGGG[C/G]CTCACAGGATGGTGCATTTGCCCTT
Phenotype
MIM: 613201 MIM: 607298
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CHTF18 PubMed Links

Gene Details

Gene
CHTF18
Gene Name
chromosome transmission fidelity factor 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022092.2 308 Intron NP_071375.1
XM_005255470.1 308 Intron XP_005255527.1
XM_005255471.3 308 Intron XP_005255528.1
XM_011522572.1 308 Intron XP_011520874.1
XM_011522573.1 308 Intron XP_011520875.1
XM_017023532.1 308 Intron XP_016879021.1
XM_017023533.1 308 Intron XP_016879022.1
XM_017023534.1 308 Intron XP_016879023.1
Gene
GNG13
Gene Name
G protein subunit gamma 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016541.2 308 UTR 3 NP_057625.1
Gene
PRR25
Gene Name
proline rich 25
There are no transcripts associated with this gene.

View Full Product Details