Product Details

SNP ID
rs147438966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:27402583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCATCAGACTGCCCCCAGCACA[C/T]GGAATGGATTTCTGAGAAAGAAGCC
Phenotype
MIM: 605383
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL21R PubMed Links

Gene Details

Gene
IL21R
Gene Name
interleukin 21 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021798.3 422 Intron NP_068570.1
NM_181078.2 422 UTR 5 NP_851564.1
NM_181079.4 422 UTR 5 NP_851565.4
XM_011545857.2 422 UTR 5 XP_011544159.1
XM_011545858.2 422 UTR 5 XP_011544160.1
XM_017023257.1 422 UTR 5 XP_016878746.1

View Full Product Details