Product Details

SNP ID
rs147758502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56962071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCACGAGGCAGGCATCGTGTGCC[A/G]CATCACCAAGCCTGCCCTCCTGGTG
Phenotype
MIM: 118470
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CETP PubMed Links

Gene Details

Gene
CETP
Gene Name
cholesteryl ester transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000078.2 109 Missense Mutation CAC,CGC H31R NP_000069.2
NM_001286085.1 109 Missense Mutation CAC,CGC H31R NP_001273014.1
XM_006721124.3 109 Missense Mutation CAC,CGC H31R XP_006721187.1

View Full Product Details