Product Details

SNP ID
rs148320997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66379340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGATCTGTTCCTCCTGGGAAGAT[A/G]CAGAGGCTCATGATGCTCCTCGCCA
Phenotype
MIM: 601120
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDH5 PubMed Links

Gene Details

Gene
CDH5
Gene Name
cadherin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001795.4 151 Missense Mutation ATA,ATG I1M NP_001786.2
XM_011522801.1 151 Missense Mutation ATA,ATG I1M XP_011521103.1

View Full Product Details