Product Details

SNP ID
rs148793965
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:15407831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTTCCCAATTCCAGAGTGGAC[C/T]GATGTACTGGCCCTTTGTACAGGTA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MPV17L PubMed Links
Additional Information
For this assay, SNP(s) [rs542512244] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MPV17L
Gene Name
MPV17 mitochondrial inner membrane protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128423.1 533 Missense Mutation CCG,CTG P130L NP_001121895.1
NM_173803.3 533 Silent Mutation ACC,ACT T106T NP_776164.2
XM_017023108.1 533 Intron XP_016878597.1

View Full Product Details