Product Details

SNP ID
rs148841919
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67930469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTGAGGCCTTCAGTCAGAGCCTC[G/T]TTTGAGGATGCCAGGCAAACTGGCG
Phenotype
MIM: 118888 MIM: 606967 MIM: 177015 MIM: 176847
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
CTRL PubMed Links
Additional Information
For this assay, SNP(s) [rs11552953] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CTRL
Gene Name
chymotrypsin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001907.2 461 Silent Mutation NP_001898.1
Gene
LCAT
Gene Name
lecithin-cholesterol acyltransferase
There are no transcripts associated with this gene.

Gene
PSKH1
Gene Name
protein serine kinase H1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006742.2 461 Intron NP_006733.1
Gene
PSMB10
Gene Name
proteasome subunit beta 10
There are no transcripts associated with this gene.

View Full Product Details