Product Details

SNP ID
rs148933926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:77202627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATTGGGAAATTAATGTATAGCCT[A/G]GGTATTAGGAGATGTTACCTGATGA
Phenotype
MIM: 608954
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MON1B PubMed Links

Gene Details

Gene
MON1B
Gene Name
MON1 homolog B, secretory trafficking associated
There are no transcripts associated with this gene.

Gene
SYCE1L
Gene Name
synaptonemal complex central element protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129979.1 Intron NP_001123451.1

View Full Product Details