Product Details

SNP ID
rs149537551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:27405918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCTGCTGATCTCTGCAGCCCA[C/T]AGGAAGGATCTGGCCACCCCGACCC
Phenotype
MIM: 605383
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL21R PubMed Links

Gene Details

Gene
IL21R
Gene Name
interleukin 21 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021798.3 Intron NP_068570.1
NM_181078.2 Intron NP_851564.1
NM_181079.4 Intron NP_851565.4
XM_011545857.2 Intron XP_011544159.1
XM_011545858.2 Intron XP_011544160.1
XM_017023257.1 Intron XP_016878746.1

View Full Product Details