Product Details

SNP ID
rs149805239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1945606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCCACGGCTTGGCGACTGTGAT[A/G]CACCAGGAGGGACTGGGGAATCCAT
Phenotype
MIM: 606216
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MSRB1 PubMed Links

Gene Details

Gene
MSRB1
Gene Name
methionine sulfoxide reductase B1
There are no transcripts associated with this gene.

Gene
RPL3L
Gene Name
ribosomal protein L3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005061.2 1567 Missense Mutation CAT,TAT H354Y NP_005052.1
XM_011522571.2 1567 Missense Mutation CAT,TAT H359Y XP_011520873.1

View Full Product Details