Product Details

SNP ID
rs149860087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4192491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTAAAGACCAGTTCTCCATCAC[C/T]GAAGAAGGTCATTTTGTCCTTGTAA
Phenotype
MIM: 604992
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRL PubMed Links

Gene Details

Gene
SRL
Gene Name
sarcalumenin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098814.1 1143 Missense Mutation AGT,GGT S362G NP_001092284.1
NM_001323667.1 1143 Missense Mutation AGT,GGT S320G NP_001310596.1
NM_001323668.1 1143 Missense Mutation AGT,GGT S320G NP_001310597.1
XM_017023527.1 1143 Missense Mutation AGT,GGT S746G XP_016879016.1
XM_017023528.1 1143 Missense Mutation AGT,GGT S320G XP_016879017.1
XM_017023529.1 1143 Missense Mutation AGT,GGT S320G XP_016879018.1

View Full Product Details