Product Details

SNP ID
rs149953957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:8894551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGGGGGGGGGAGAACCCTTACC[A/G]GGCTGTGGCTCAAAGTCTTTCAAAT
Phenotype
MIM: 602519
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929989 PubMed Links

Gene Details

Gene
LOC101929989
Gene Name
lipopolysaccharide-induced tumor necrosis factor-alpha factor-like
There are no transcripts associated with this gene.

Gene
USP7
Gene Name
ubiquitin specific peptidase 7 (herpes virus-associated)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286457.1 3400 Silent Mutation CCC,CCT P1051P NP_001273386.1
NM_001286458.1 3400 Silent Mutation CCC,CCT P968P NP_001273387.1
NM_001321858.1 3400 Silent Mutation CCC,CCT P1009P NP_001308787.1
NM_003470.2 3400 Silent Mutation CCC,CCT P1067P NP_003461.2
XM_017023652.1 3400 Silent Mutation CCC,CCT P1067P XP_016879141.1
XM_017023653.1 3400 Silent Mutation CCC,CCT P1051P XP_016879142.1

View Full Product Details