Product Details

SNP ID
rs150184095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30666939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGAAACCAGGGAAGTGGTGTGC[C/T]ATGCACGTGCGTGTGGCTTACATGA
Phenotype
MIM: 608601
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBRS PubMed Links

Gene Details

Gene
FBRS
Gene Name
fibrosin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105079.2 3171 Silent Mutation GCC,GCT A608A NP_001098549.2
XM_011545916.2 3171 Silent Mutation GCC,GCT A598A XP_011544218.1
XM_011545917.2 3171 Silent Mutation GCC,GCT A598A XP_011544219.1
XM_011545919.2 3171 Silent Mutation GCC,GCT A586A XP_011544221.1

View Full Product Details