Product Details

SNP ID
rs150400958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3285118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGAGCCTTTGCCTCTGGAAAC[A/T]GCACGAGAGTCACCGAGCTTCAAGC
Phenotype
MIM: 604191
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF263 PubMed Links

Gene Details

Gene
ZNF263
Gene Name
zinc finger protein 263
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005741.4 886 Silent Mutation ACA,ACT T149T NP_005732.2
XM_006720831.1 886 Silent Mutation ACA,ACT T149T XP_006720894.1
XM_006720832.1 886 Silent Mutation ACA,ACT T149T XP_006720895.1
XM_006720833.1 886 Intron XP_006720896.1
XM_006720834.3 886 Intron XP_006720897.1
XM_011522344.1 886 Silent Mutation ACA,ACT T149T XP_011520646.1
XM_017022850.1 886 Intron XP_016878339.1

View Full Product Details