Product Details

SNP ID
rs150759445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4694034 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCGGGGGCTTCGTGGACCGGCG[C/T]TTCTGGTCGCTGGAGGACGGCCTGA
Phenotype
MIM: 607559
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKS3 PubMed Links

Gene Details

Gene
ANKS3
Gene Name
ankyrin repeat and sterile alpha motif domain containing 3
There are no transcripts associated with this gene.

Gene
MGRN1
Gene Name
mahogunin ring finger 1
There are no transcripts associated with this gene.

Gene
NUDT16L1
Gene Name
nudix hydrolase 16 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193452.1 244 Silent Mutation CGC,CGT R70R NP_001180381.1
NM_032349.3 244 Silent Mutation CGC,CGT R70R NP_115725.1
XM_005255633.4 244 Silent Mutation CGC,CGT R84R XP_005255690.1

View Full Product Details