Product Details

SNP ID
rs150789330
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31436847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCAGGGCAAAGGGGAGGCTTCT[C/T]ATGAGCAGGGCTTCGGAGATGACCG
Phenotype
MIM: 602009
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
COX6A2 PubMed Links

Gene Details

Gene
COX6A2
Gene Name
cytochrome c oxidase subunit 6A2
There are no transcripts associated with this gene.

Gene
ZNF843
Gene Name
zinc finger protein 843
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136509.1 730 Missense Mutation ATA,ATG I0M NP_001129981.1

View Full Product Details