Product Details

SNP ID
rs151006109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88653352 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGGGGGTCGGCTCCATGGCCAGC[A/G]CAGCCTGAAGCTCAGCTGAGAGAGG
Phenotype
MIM: 608508 MIM: 603236
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYBA PubMed Links

Gene Details

Gene
CYBA
Gene Name
cytochrome b-245 alpha chain
There are no transcripts associated with this gene.

Gene
MVD
Gene Name
mevalonate diphosphate decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002461.2 1245 Missense Mutation GCG,GTG A357V NP_002452.1
XM_011523086.2 1245 Missense Mutation GCG,GTG A391V XP_011521388.1
XM_011523087.2 1245 Missense Mutation GCG,GTG A370V XP_011521389.1
XM_011523088.2 1245 Missense Mutation GCG,GTG A336V XP_011521390.1
XM_011523089.2 1245 Missense Mutation GCG,GTG A240V XP_011521391.1

View Full Product Details