Product Details

SNP ID
rs151189567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20553813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGCCTTGAGGCCCAGGCTCGAG[C/T]AGGAATGTTCTGCCATCTTGGGAAG
Phenotype
MIM: 614359
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACSM2B PubMed Links

Gene Details

Gene
ACSM2B
Gene Name
acyl-CoA synthetase medium-chain family member 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105069.1 Intron NP_001098539.1
NM_182617.3 Intron NP_872423.3
XM_005255304.3 Intron XP_005255361.1
XM_011545824.2 Intron XP_011544126.1
XM_017023205.1 Intron XP_016878694.1

View Full Product Details