Product Details

SNP ID
rs151206219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70156495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAGGGCATGGATTTCATTGGTCG[C/T]GACGCCCTCCTGCAGCAGAAGCAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC400541 PubMed Links

Gene Details

Gene
LOC400541
Gene Name
uncharacterized LOC400541
There are no transcripts associated with this gene.

Gene
PDPR
Gene Name
pyruvate dehydrogenase phosphatase regulatory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322117.1 2863 Silent Mutation CGC,CGT R752R NP_001309046.1
NM_001322118.1 2863 Silent Mutation CGC,CGT R652R NP_001309047.1
NM_001322119.1 2863 Silent Mutation CGC,CGT R350R NP_001309048.1
NM_017990.4 2863 Silent Mutation CGC,CGT R752R NP_060460.4
XM_005256015.2 2863 Silent Mutation CGC,CGT R752R XP_005256072.1
XM_005256016.3 2863 Silent Mutation CGC,CGT R752R XP_005256073.1
XM_006721224.2 2863 Silent Mutation CGC,CGT R691R XP_006721287.1
XM_006721225.3 2863 Intron XP_006721288.1
XM_011523190.2 2863 Intron XP_011521492.1
XM_017023387.1 2863 Silent Mutation CGC,CGT R752R XP_016878876.1
XM_017023388.1 2863 Intron XP_016878877.1
XM_017023389.1 2863 Intron XP_016878878.1

View Full Product Details