Product Details

SNP ID
rs151318611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69339548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCGATGGGATAGTCGCCGCGGTC[A/G]CCATCTTCCCAGCAACAACGTCACT
Phenotype
MIM: 606979
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COG8 PubMed Links

Gene Details

Gene
COG8
Gene Name
component of oligomeric golgi complex 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032382.4 37 Missense Mutation GCG,GTG A2V NP_115758.3
Gene
NIP7
Gene Name
NIP7, nucleolar pre-rRNA processing protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199434.1 37 UTR 5 NP_001186363.1
NM_016101.4 37 UTR 5 NP_057185.1
Gene
PDF
Gene Name
peptide deformylase (mitochondrial)
There are no transcripts associated with this gene.

Gene
TMED6
Gene Name
transmembrane p24 trafficking protein 6
There are no transcripts associated with this gene.

View Full Product Details