Product Details

SNP ID
rs137989863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81912442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACGTCACTTTCTTCCTTTTTGTGC[A/G]TTTTGTGCAGCCCCTGCCAAACCAG
Phenotype
MIM: 602020 MIM: 607996 MIM: 602679 MIM: 606212
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAFG PubMed Links

Gene Details

Gene
MAFG
Gene Name
MAF bZIP transcription factor G
There are no transcripts associated with this gene.

Gene
NPB
Gene Name
neuropeptide B
There are no transcripts associated with this gene.

Gene
PCYT2
Gene Name
phosphate cytidylyltransferase 2, ethanolamine
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184917.2 2831 Intron NP_001171846.1
NM_001256433.2 2831 Intron NP_001243362.1
NM_001256434.2 2831 Intron NP_001243363.1
NM_001256435.2 2831 Intron NP_001243364.1
NM_001282203.1 2831 Intron NP_001269132.1
NM_001282204.1 2831 Intron NP_001269133.1
NM_002861.4 2831 Intron NP_002852.1
XM_005256386.3 2831 Intron XP_005256443.1
XM_005256387.3 2831 Intron XP_005256444.1
XM_006722287.3 2831 Intron XP_006722350.1
XM_017024910.1 2831 Intron XP_016880399.1
XM_017024911.1 2831 Intron XP_016880400.1
XM_017024912.1 2831 Intron XP_016880401.1
Gene
SIRT7
Gene Name
sirtuin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016538.2 2831 Missense Mutation CGC,TGC R393C NP_057622.1
XM_006722284.2 2831 Missense Mutation CGC,TGC R311C XP_006722347.1
XM_011523580.1 2831 Missense Mutation CGC,TGC R358C XP_011521882.1
XM_011523581.2 2831 Missense Mutation CGC,TGC R322C XP_011521883.1

View Full Product Details