Product Details

SNP ID
rs138158637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48577928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTCTTTGCACGCGGAGTGGGAC[A/G]GGCTGGGGTGCAGGGGGTTCTGGGC
Phenotype
MIM: 142966 MIM: 142965 MIM: 610173
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOXB3 PubMed Links

Gene Details

Gene
HOXB3
Gene Name
homeobox B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002146.4 454 Intron NP_002137.4
XM_005257277.3 454 Intron XP_005257334.1
XM_005257280.3 454 Intron XP_005257337.1
XM_005257282.3 454 Intron XP_005257339.1
XM_006721854.2 454 Intron XP_006721917.1
XM_011524707.2 454 Intron XP_011523009.1
XM_011524708.2 454 Intron XP_011523010.1
XM_011524710.1 454 Intron XP_011523012.1
XM_011524719.1 454 Intron XP_011523021.1
XM_011524720.2 454 Intron XP_011523022.1
XM_011524721.2 454 Intron XP_011523023.1
XM_011524726.2 454 Intron XP_011523028.1
XM_017024557.1 454 Intron XP_016880046.1
XM_017024558.1 454 Intron XP_016880047.1
XM_017024559.1 454 Intron XP_016880048.1
XM_017024560.1 454 Intron XP_016880049.1
XM_017024561.1 454 Intron XP_016880050.1
XM_017024562.1 454 Intron XP_016880051.1
XM_017024563.1 454 Intron XP_016880052.1
Gene
HOXB4
Gene Name
homeobox B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024015.4 454 Missense Mutation CCG,CTG P131L NP_076920.1
Gene
MIR10A
Gene Name
microRNA 10a
There are no transcripts associated with this gene.

View Full Product Details