Product Details
- SNP ID
-
rs138388461
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:59169823 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAGATTATTCAAAAAAAAAGAAG[C/G]CTCTCACTTTCAGTCCAAGCTAATT
- Phenotype
-
MIM: 615920
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PRR11
PubMed Links
Gene Details
- Gene
- PRR11
- Gene Name
- proline rich 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018304.3 |
383 |
Missense Mutation |
GCC,GGC |
A24G |
NP_060774.2 |
View Full Product Details