Product Details
- SNP ID
-
rs138601922
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:63524223 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACGGCTTCTGCGAACTCTTCGGCT[A/G]CTCCCGAGTGGAGGTGATGCAGCAA
- Phenotype
-
MIM: 608168
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KCNH6
PubMed Links
Gene Details
- Gene
- KCNH6
- Gene Name
- potassium voltage-gated channel subfamily H member 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001278919.1 |
241 |
Missense Mutation |
TAC,TGC |
Y54C |
NP_001265848.1 |
NM_001278920.1 |
241 |
UTR 5 |
|
|
NP_001265849.1 |
NM_030779.3 |
241 |
Missense Mutation |
TAC,TGC |
Y54C |
NP_110406.1 |
NM_173092.2 |
241 |
Missense Mutation |
TAC,TGC |
Y54C |
NP_775115.1 |
XM_011525308.2 |
241 |
Missense Mutation |
TAC,TGC |
Y54C |
XP_011523610.1 |
XM_011525309.2 |
241 |
Missense Mutation |
TAC,TGC |
Y54C |
XP_011523611.1 |
XM_011525310.2 |
241 |
Missense Mutation |
TAC,TGC |
Y54C |
XP_011523612.1 |
XM_011525311.1 |
241 |
Missense Mutation |
TAC,TGC |
Y54C |
XP_011523613.1 |
XM_011525312.1 |
241 |
Missense Mutation |
TAC,TGC |
Y54C |
XP_011523614.1 |
XM_011525313.1 |
241 |
Missense Mutation |
TAC,TGC |
Y54C |
XP_011523615.1 |
XM_017025175.1 |
241 |
Missense Mutation |
TAC,TGC |
Y54C |
XP_016880664.1 |
XM_017025176.1 |
241 |
Missense Mutation |
TAC,TGC |
Y54C |
XP_016880665.1 |
XM_017025177.1 |
241 |
Missense Mutation |
TAC,TGC |
Y54C |
XP_016880666.1 |
XM_017025178.1 |
241 |
Missense Mutation |
TAC,TGC |
Y54C |
XP_016880667.1 |
XM_017025179.1 |
241 |
Missense Mutation |
TAC,TGC |
Y54C |
XP_016880668.1 |
XM_017025180.1 |
241 |
UTR 5 |
|
|
XP_016880669.1 |
View Full Product Details