Product Details

SNP ID
rs138622503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44014362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGGGCCGAGGCGCTCACCTTCA[A/G]CCAGTCCCCGTAGTGGACGTAGCCC
Phenotype
MIM: 608300 MIM: 600781
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAGS PubMed Links

Gene Details

Gene
NAGS
Gene Name
N-acetylglutamate synthase
There are no transcripts associated with this gene.

Gene
PYY
Gene Name
peptide YY
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004160.5 558 Intron NP_004151.3
XM_011525035.1 558 Intron XP_011523337.1
Gene
TMEM101
Gene Name
transmembrane protein 101
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304813.1 558 Silent Mutation NP_001291742.1
NM_001304814.1 558 Silent Mutation NP_001291743.1
NM_032376.3 558 Silent Mutation NP_115752.1
XM_005257737.2 558 Silent Mutation XP_005257794.1
XM_011525346.1 558 Silent Mutation XP_011523648.1
XM_011525347.1 558 Silent Mutation XP_011523649.1
XM_011525349.1 558 Silent Mutation XP_011523651.1
XM_011525351.1 558 Silent Mutation XP_011523653.1
XM_011525353.1 558 Silent Mutation XP_011523655.1

View Full Product Details