Product Details

SNP ID
rs138782271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7173716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCTCTGTCTCGCAGACCCAGCG[A/G]TAGGGCCTCTGGCAGACGTCGTCGT
Phenotype
MIM: 108360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASGR1 PubMed Links

Gene Details

Gene
ASGR1
Gene Name
asialoglycoprotein receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197216.2 1126 Silent Mutation TAC,TAT Y234Y NP_001184145.1
NM_001671.4 1126 Silent Mutation TAC,TAT Y273Y NP_001662.1
XM_011523861.2 1126 Silent Mutation TAC,TAT Y273Y XP_011522163.1

View Full Product Details