Product Details

SNP ID
rs139119751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4797483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTGACAAGCTGACACCTATTCA[C/T]GACCGCATTTTCTGCTGTCGCTCAG
Phenotype
MIM: 602384 MIM: 600307
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLTPD2 PubMed Links

Gene Details

Gene
GLTPD2
Gene Name
glycolipid transfer protein domain containing 2
There are no transcripts associated with this gene.

Gene
LOC107984973
Gene Name
atherin-like
There are no transcripts associated with this gene.

Gene
PLD2
Gene Name
phospholipase D2
There are no transcripts associated with this gene.

Gene
PSMB6
Gene Name
proteasome subunit beta 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270481.1 267 Silent Mutation CAC,CAT H72H NP_001257410.1
NM_002798.2 267 Silent Mutation CAC,CAT H72H NP_002789.1

View Full Product Details