Product Details

SNP ID
rs139277198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76391155 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGTGGCACCGATGCGTCTGGTG[C/T]GGTCTCCGAAGTCTGGTCTGTGTGG
Phenotype
MIM: 603730
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPHK1 PubMed Links

Gene Details

Gene
SPHK1
Gene Name
sphingosine kinase 1
There are no transcripts associated with this gene.

Gene
UBE2O
Gene Name
ubiquitin conjugating enzyme E2 O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022066.3 3641 Missense Mutation ACA,GCA T1223A NP_071349.3
XM_005257567.1 3641 Missense Mutation ACA,GCA T1224A XP_005257624.1
XM_017024934.1 3641 Missense Mutation ACA,GCA T1199A XP_016880423.1
XM_017024935.1 3641 Missense Mutation ACA,GCA T1198A XP_016880424.1

View Full Product Details