Product Details

SNP ID
rs139339869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81510933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAACCCCTCGACTCACCTTGAT[C/T]TTCATGGTGCTGGGCGCCAGGGCGG
Phenotype
MIM: 102560 MIM: 607643
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTG1 PubMed Links

Gene Details

Gene
ACTG1
Gene Name
actin gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199954.1 1117 Silent Mutation AAA,AAG K326K NP_001186883.1
NM_001614.3 1117 Silent Mutation AAA,AAG K326K NP_001605.1
Gene
FSCN2
Gene Name
fascin actin-bundling protein 2, retinal
There are no transcripts associated with this gene.

View Full Product Details