Product Details

SNP ID
rs139456598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44397571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTAGGATCCCATCCCCCAACTCA[C/T]GTGCCATGGCTGGGGGGATGTTGCA
Phenotype
MIM: 614396 MIM: 607759
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPATCH8 PubMed Links

Gene Details

Gene
GPATCH8
Gene Name
G-patch domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002909.3 4605 Silent Mutation NP_001002909.1
NM_001304939.1 4605 Silent Mutation NP_001291868.1
NM_001304940.1 4605 Silent Mutation NP_001291869.1
NM_001304941.1 4605 Silent Mutation NP_001291870.1
NM_001304942.1 4605 Silent Mutation NP_001291871.1
NM_001304943.1 4605 Silent Mutation NP_001291872.1
XM_011524557.1 4605 Silent Mutation XP_011522859.1
XM_011524558.1 4605 Silent Mutation XP_011522860.1
XM_011524559.2 4605 Silent Mutation XP_011522861.1
XM_011524561.2 4605 Silent Mutation XP_011522863.1
XM_017024372.1 4605 Silent Mutation XP_016879861.1
XM_017024373.1 4605 Silent Mutation XP_016879862.1
XM_017024374.1 4605 Silent Mutation XP_016879863.1
XM_017024375.1 4605 Silent Mutation XP_016879864.1
XM_017024376.1 4605 Silent Mutation XP_016879865.1
XM_017024377.1 4605 Silent Mutation XP_016879866.1
XM_017024378.1 4605 Silent Mutation XP_016879867.1
XM_017024379.1 4605 Silent Mutation XP_016879868.1
XM_017024380.1 4605 Silent Mutation XP_016879869.1
Gene
ITGA2B
Gene Name
integrin subunit alpha 2b
There are no transcripts associated with this gene.

View Full Product Details