Product Details

SNP ID
rs139746240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4632048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTGCATTAGGCACCCAAGAGTA[C/T]CAGTCCAGCTAAGGAAGGGCAGGGA
Phenotype
MIM: 152392
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALOX15 PubMed Links

Gene Details

Gene
ALOX15
Gene Name
arachidonate 15-lipoxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001140.3 1664 Nonsense Mutation NP_001131.3

View Full Product Details