Product Details

SNP ID
rs139897506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74919578 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCGGAGGTCGAGGTCAGAGCACA[C/G]CATCAAAGCCTCGAGGTCGATCTTC
Phenotype
MIM: 607827 MIM: 607696
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OTOP2 PubMed Links

Gene Details

Gene
OTOP2
Gene Name
otopetrin 2
There are no transcripts associated with this gene.

Gene
USH1G
Gene Name
USH1 protein network component sans
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282489.2 1421 Missense Mutation CTG,GTG L317V NP_001269418.1
NM_173477.4 1421 Missense Mutation CTG,GTG L420V NP_775748.2
XM_011524296.2 1421 Missense Mutation CTG,GTG L317V XP_011522598.1

View Full Product Details