Product Details

SNP ID
rs139907815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44005822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCAAGAGCTGCAAGGTGCTGGT[A/G]GACGCGCTTCGACACAACGCCGCCG
Phenotype
MIM: 608300 MIM: 600781
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAGS PubMed Links

Gene Details

Gene
NAGS
Gene Name
N-acetylglutamate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153006.2 502 Silent Mutation GTA,GTG V204V NP_694551.1
XM_011524438.1 502 Silent Mutation GTA,GTG V204V XP_011522740.1
XM_011524439.2 502 Silent Mutation GTA,GTG V38V XP_011522741.1
Gene
PYY
Gene Name
peptide YY
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004160.5 502 Intron NP_004151.3
XM_011525035.1 502 Intron XP_011523337.1
Gene
TMEM101
Gene Name
transmembrane protein 101
There are no transcripts associated with this gene.

View Full Product Details