Product Details

SNP ID
rs140119129
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:56981197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATGCCACCAACTCCTGCAAGAA[A/C]TTCTCAGAACTGCCCCTGGTCATGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SCPEP1 PubMed Links

Gene Details

Gene
SCPEP1
Gene Name
serine carboxypeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021626.2 227 Missense Mutation AAA,AAC K64N NP_067639.1
XM_005257557.2 227 Missense Mutation AAA,AAC K64N XP_005257614.1

View Full Product Details