Product Details
- SNP ID
-
rs140506773
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:43877824 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACAGGCTCAGTACACCCAGCTGA[C/G]AGGCACCCACTGGGGCTCTGTCCGT
- Phenotype
-
MIM: 600723
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MPP2
PubMed Links
Gene Details
- Gene
- MPP2
- Gene Name
- membrane palmitoylated protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001278370.1 |
1890 |
Missense Mutation |
CTC,GTC |
L593V |
NP_001265299.1 |
NM_001278371.1 |
1890 |
Missense Mutation |
CTC,GTC |
L537V |
NP_001265300.1 |
NM_001278372.1 |
1890 |
Missense Mutation |
CTC,GTC |
L572V |
NP_001265301.1 |
NM_001278373.1 |
1890 |
Missense Mutation |
CTC,GTC |
L537V |
NP_001265302.1 |
NM_001278374.1 |
1890 |
Missense Mutation |
CTC,GTC |
L409V |
NP_001265303.1 |
NM_001278375.1 |
1890 |
Missense Mutation |
CTC,GTC |
L537V |
NP_001265304.1 |
NM_001278376.2 |
1890 |
Missense Mutation |
CTC,GTC |
L565V |
NP_001265305.2 |
NM_001278381.1 |
1890 |
Missense Mutation |
CTC,GTC |
L548V |
NP_001265310.1 |
NM_005374.4 |
1890 |
Missense Mutation |
CTC,GTC |
L548V |
NP_005365.4 |
XM_011524827.2 |
1890 |
Missense Mutation |
CTC,GTC |
L626V |
XP_011523129.2 |
View Full Product Details