Product Details

SNP ID
rs141056088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47744293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGCCTGCAACGCTTCCAACAC[G/T]CATATCTTTACTTTCCAAGAAACCC
Phenotype
MIM: 604895
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TBX21 PubMed Links

Gene Details

Gene
TBX21
Gene Name
T-box 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013351.1 1145 Silent Mutation ACG,ACT T289T NP_037483.1
XM_011524698.2 1145 Silent Mutation ACG,ACT T310T XP_011523000.1

View Full Product Details