Product Details
- SNP ID
-
rs141165409
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:75898879 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGATGCCATAGGTGGGCTCATGAC[A/T]GTCCCTGTACCGGTCCAGGTAGCGC
- Phenotype
-
MIM: 611844
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MRPL38
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9191] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MRPL38
- Gene Name
- mitochondrial ribosomal protein L38
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032478.3 |
1339 |
Missense Mutation |
AGT,TGT |
S372C |
NP_115867.2 |
- Gene
- TRIM65
- Gene Name
- tripartite motif containing 65
There are no transcripts associated with this gene.
View Full Product Details