Product Details
- SNP ID
-
rs141306639
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:74863943 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCACATTGGGGATGACCCCAAGC[C/T]TGGAGTCAAAGGGCACGCTTGGGTC
- Phenotype
-
MIM: 103270
MIM: 138254
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FDXR
PubMed Links
Gene Details
- Gene
- FDXR
- Gene Name
- ferredoxin reductase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001258012.3 |
1272 |
Missense Mutation |
AAG,AGG |
K419R |
NP_001244941.2 |
NM_001258013.3 |
1272 |
Missense Mutation |
AAG,AGG |
K407R |
NP_001244942.2 |
NM_001258014.3 |
1272 |
Missense Mutation |
AAG,AGG |
K368R |
NP_001244943.2 |
NM_001258015.2 |
1272 |
Missense Mutation |
AAG,AGG |
K336R |
NP_001244944.1 |
NM_001258016.3 |
1272 |
Missense Mutation |
AAG,AGG |
K324R |
NP_001244945.2 |
NM_004110.5 |
1272 |
Missense Mutation |
AAG,AGG |
K382R |
NP_004101.3 |
NM_024417.4 |
1272 |
Missense Mutation |
AAG,AGG |
K376R |
NP_077728.3 |
XM_006721772.1 |
1272 |
Missense Mutation |
AAG,AGG |
K377R |
XP_006721835.1 |
XM_011524527.1 |
1272 |
Missense Mutation |
AAG,AGG |
K383R |
XP_011522829.1 |
XM_011524528.1 |
1272 |
Missense Mutation |
AAG,AGG |
K380R |
XP_011522830.1 |
XM_011524529.1 |
1272 |
Missense Mutation |
AAG,AGG |
K379R |
XP_011522831.1 |
XM_011524531.2 |
1272 |
Missense Mutation |
AAG,AGG |
K331R |
XP_011522833.1 |
XM_011524532.2 |
1272 |
Missense Mutation |
AAG,AGG |
K330R |
XP_011522834.1 |
XM_011524533.2 |
1272 |
Missense Mutation |
AAG,AGG |
K325R |
XP_011522835.1 |
- Gene
- GRIN2C
- Gene Name
- glutamate ionotropic receptor NMDA type subunit 2C
There are no transcripts associated with this gene.
View Full Product Details