Product Details

SNP ID
rs141393724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64150866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAACTCTTGCTCCAGTTTCTTC[C/T]CTATCCAGTCTGTCACATGAACTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNHG25 PubMed Links

Gene Details

Gene
SNHG25
Gene Name
small nucleolar RNA host gene 25
There are no transcripts associated with this gene.

Gene
SNORA50C
Gene Name
small nucleolar RNA, H/ACA box 50C
There are no transcripts associated with this gene.

Gene
SNORD104
Gene Name
small nucleolar RNA, C/D box 104
There are no transcripts associated with this gene.

Gene
TEX2
Gene Name
testis expressed 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288732.1 3381 Missense Mutation NP_001275661.1
NM_001288733.1 3381 Missense Mutation NP_001275662.1
NM_018469.4 3381 Missense Mutation NP_060939.3
XM_011524998.1 3381 Missense Mutation XP_011523300.1
XM_011524999.1 3381 Missense Mutation XP_011523301.1
XM_011525000.1 3381 Missense Mutation XP_011523302.1
XM_017024846.1 3381 Missense Mutation XP_016880335.1
XM_017024847.1 3381 Intron XP_016880336.1

View Full Product Details