Product Details

SNP ID
rs141415687
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42295698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAACCAGACATTTGAGGAGCTGC[A/G]ACTGGTCACGCAGGACACAGAGAAT
Phenotype
MIM: 601511
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STAT5A PubMed Links

Gene Details

Gene
STAT5A
Gene Name
signal transducer and activator of transcription 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288718.1 715 Missense Mutation CAA,CGA Q152R NP_001275647.1
NM_001288719.1 715 Missense Mutation CAA,CGA Q122R NP_001275648.1
NM_001288720.1 715 Missense Mutation CAA,CGA Q152R NP_001275649.1
NM_003152.3 715 Missense Mutation CAA,CGA Q152R NP_003143.2
XM_005257624.3 715 Missense Mutation CAA,CGA Q152R XP_005257681.1

View Full Product Details