Product Details

SNP ID
rs141526407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81406594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCGCACTCCGCTCGGGCTCCGCC[C/T]GCTGACGTTTTTATGGGATAATATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAHCC1 PubMed Links

Gene Details

Gene
BAHCC1
Gene Name
BAH domain and coiled-coil containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291324.1 Intron NP_001278253.1
XM_011525063.2 Intron XP_011523365.1
XM_017024898.1 Intron XP_016880387.1
XM_017024899.1 Intron XP_016880388.1
Gene
MIR4740
Gene Name
microRNA 4740
There are no transcripts associated with this gene.

View Full Product Details