Product Details

SNP ID
rs141638018
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18018299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAATATTTAATAAAATCAGAGTC[A/G]AGAGAAAGTCACTTGCACAATTCAT
Phenotype
MIM: 608918
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ATPAF2 PubMed Links

Gene Details

Gene
ATPAF2
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145691.3 1129 UTR 3 NP_663729.1
XM_005256848.3 1129 Intron XP_005256905.1
XM_011524065.1 1129 Intron XP_011522367.1
XM_017025302.1 1129 UTR 3 XP_016880791.1
XM_017025303.1 1129 Intron XP_016880792.1
Gene
DRC3
Gene Name
dynein regulatory complex subunit 3
There are no transcripts associated with this gene.

View Full Product Details